missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human RNF170 Partial ORF (NP_112216, 121 a.a. - 193 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00081790-Q01.10ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Sequence: LGAISCPICRQTVTLLLTVFGEDDQSQDVLRLHQDINDYNRRFSGQPRSIMERIMDLPTLLRHAFREMFSVGGEspecificaciones
NP_112216 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
33.77kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LGAISCPICRQTVTLLLTVFGEDDQSQDVLRLHQDINDYNRRFSGQPRSIMERIMDLPTLLRHAFREMFSVGG | |
RUO | |
RNF170 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
81790 | |
RNF170 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp564A022/FLJ38306 | |
RNF170 | |
Recombinant | |
wheat germ expression system |