missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human RNF146 Full-length ORF (NP_112225.2, 1 a.a. - 358 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00081847-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: MAGCGEIDHSINMLPTNRKANESCSNTAPSLTVPECAICLQTCVHPVSLPCKHVFCYLCVKGASWLGKRCALCRQEIPEDFLDKPTLLSPEELKAASRGNGEYAWYYEGRNGWWQYDERTSRELEDAFSKGKKNTEMLIAGFLYVADLENMVQYRRNEHGRRRKIKRDIIDIPKKGVAGLRLDCDANTVNLARESSADGADSVSAQSGASVQPLVSSVRPLTSVDGQLTSPATPSPDASTSLEDSFAHLQLSGDNTAERSHRGEGEEDHESPSSGRVPAPDTSIEETESDASSDSEDVSAVVAQHSLTQQRLLVSNANQTVPDRSDRSGTDRSVAGGGTVSVSVRSRRPDGQCTVTEVEspecificaciones
NP_112225.2 | |
Liquid | |
81847 | |
RNF146 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAGCGEIDHSINMLPTNRKANESCSNTAPSLTVPECAICLQTCVHPVSLPCKHVFCYLCVKGASWLGKRCALCRQEIPEDFLDKPTLLSPEELKAASRGNGEYAWYYEGRNGWWQYDERTSRELEDAFSKGKKNTEMLIAGFLYVADLENMVQYRRNEHGRRRKIKRDIIDIPKKGVAGLRLDCDANTVNLARESSADGADSVSAQSGASVQPLVSSVRPLTSVDGQLTSPATPSPDASTSLEDSFAHLQLSGDNTAERSHRGEGEEDHESPSSGRVPAPDTSIEETESDASSDSEDVSAVVAQHSLTQQRLLVSNANQTVPDRSDRSGTDRSVAGGGTVSVSVRSRRPDGQCTVTEV | |
RUO | |
RNF146 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
65.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp434O1427/RP3-351K20.1/dJ351K20.1 | |
RNF146 | |
Yes | |
wheat germ expression system |