missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human RGS11 Partial ORF (NP_003825, 101 a.a. - 199 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Especificaciones
Número de acceso | NP_003825 |
---|---|
Para utilizar con (aplicación) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulación | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
ID de gen (Entrez) | 8786 |
Peso molecular | 36.63kDa |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
16179245
|
Abnova™
H00008786-Q01.25ug |
25 ug |
508.00€
25 microgramos |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
16169245
|
Abnova™
H00008786-Q01.10ug |
10 ug |
335.00€
10 microgramos |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
Description
The protein encoded by this gene belongs to the RGS (regulator of G protein signaling) family. Members of the RGS family act as GTPase-activating proteins on the alpha subunits of heterotrimeric, signal-transducing G proteins. This protein inhibits signal transduction by increasing the GTPase activity of G protein alpha subunits, thereby driving them into their inactive GDP-bound form. Alternative splicing occurs at this locus and two transcript variants encoding distinct isoforms have been identified. [provided by RefSeq]
Sequence: DYAIYLAKKNIRKRGTLVDYEKDCYDRLHKKINHAWDLVLMQAREQLRAAKQRSKGDRLVIACQEQTYWLVNRPPPGAPDVLEQGPGRGSCAASRVLMTSpecifications
NP_003825 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
RS11 | |
RGS11 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
8786 | |
RGS11 (Human) Recombinant Protein (Q01) | |
DYAIYLAKKNIRKRGTLVDYEKDCYDRLHKKINHAWDLVLMQAREQLRAAKQRSKGDRLVIACQEQTYWLVNRPPPGAPDVLEQGPGRGSCAASRVLMT | |
RUO | |
RGS11 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |