missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human RBM14 Partial ORF (AAH00488, 51 a.a. - 160 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00010432-Q01.25ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Sequence: AIEALHGHELRPGRALVVEMSRPRPLNTWKIFVGNVSAACTSQELRSLFERRGRVIECDVVKDYAFVHMEKEADAKAAIAQLNGKEVKGKRINVELSTKGQKKGPGLAVQEspecificaciones
AAH00488 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
AIEALHGHELRPGRALVVEMSRPRPLNTWKIFVGNVSAACTSQELRSLFERRGRVIECDVVKDYAFVHMEKEADAKAAIAQLNGKEVKGKRINVELSTKGQKKGPGLAVQ | |
RUO | |
RBM14 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
10432 | |
RBM14 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
COAA/DKFZp779J0927/MGC15912/MGC31756/PSP2/SIP/SYTIP1/TMEM137 | |
RBM14 | |
Recombinant | |
wheat germ expression system |