missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human PTCH2 Partial ORF (NP_003729, 79 a.a. - 188 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00008643-Q01.25ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This gene encodes a member of the patched gene family. The patched protein is the receptor for sonic hedgehog, a secreted molecule implicated in the formation of embryonic structures and in tumorigenesis. This gene is shown to be mutated in a medulloblastoma and in a basal cell carcinoma, suggesting that it plays a role in the development of some tumors. Alternative transcript variants have been described, but their biological function has not been determined. [provided by RefSeq]
Sequence: ETNLEQLWVEVGSRVSQELHYTKEKLGEEAAYTSQMLIQTARQEGENILTPEALGLHLQAALTASKVQVSLYGKSWDLNKICYKSGVPLIENGMIERMIEKLFPCVILTPEspecificaciones
NP_003729 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
ETNLEQLWVEVGSRVSQELHYTKEKLGEEAAYTSQMLIQTARQEGENILTPEALGLHLQAALTASKVQVSLYGKSWDLNKICYKSGVPLIENGMIERMIEKLFPCVILTP | |
RUO | |
PTCH2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
8643 | |
PTCH2 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PTCH2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |