Learn More
ACROBiosystems Human PD-L2 / B7-DC Protein, mouse IgG1 Fc Tag
Marca: ACROBiosystems PD2-H52A5-100UG
Detalles adicionales : Peso : 0.00001kg
Descripción
Programmed cell death 1 ligand 2 (PD-L2 or PDCD1 ligand 2) is also known as Butyrophilin B7-DC, CD antigen CD273, which belongs to the immunoglobulin superfamily or BTN/MOG family. The expression of PD-L2 is up-regulated by IFNG/IFN-gamma stimulation in monocytes and induced on dendritic cells grown from peripheral blood mononuclear cells with CSF2 and IL-4. PD-L2 Involved in the costimulatory signal, essential for T-cell proliferation and IFNG production in a PDCD1-independent manner. PD-L2 interaction with PDCD1 inhibits T-cell proliferation by blocking cell cycle progression and cytokine production.
This protein carries a mouse IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 48.9 kDa. The protein migrates as 60-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
AAI13679 | |
48.9 kDa | |
100 µg | |
Leu 20 - Pro 219 | |
PDL2,PD-L2,Butyrophilin B7-DC,CD273,PDCD1 ligand 2,PDCD1L2,PDCD1LG2 | |
Unconjugated | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Human |
Powder | |
>95% as determined by reduced SDS-PAGE | |
Human PD-L2, Mouse IgG1 Fc Tag (PD2-H52A5) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Pro 219 (Accession # AAI13679). | |
-20°C | |
PD-L2 | |
Mouse IgG Fc Tag | |
Less than 1.0 EU / µg by the LAL method. | |
Immune Checkpoint Proteins |