Learn More
Abnova™ Human ORC2L Full-length ORF (AAH14834, 1 a.a. - 577 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Especificaciones
Número de acceso | AAH14834 |
---|---|
Formulación | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
ID de gen (Entrez) | 4999 |
Peso molecular | 89.21kDa |
Método de purificación | Glutathione Sepharose 4 Fast Flow |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
16118611
|
Abnova™
H00004999-P01.25ug |
25μg |
508.00€
25 microgramos |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
16108611
|
Abnova™
H00004999-P01.10ug |
10 ug |
335.00€
10 microgramos |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
Descripción
The origin recognition complex (ORC) is a highly conserved six subunits protein complex essential for the initiation of the DNA replication in eukaryotic cells. Studies in yeast demonstrated that ORC binds specifically to origins of replication and serves as a platform for the assembly of additional initiation factors such as Cdc6 and Mcm proteins. The protein encoded by this gene is a subunit of the ORC complex. This protein forms a core complex with ORC3L, -4L, and -5L. It also interacts with CDC45L and MCM10, which are proteins known to be important for the initiation of DNA replication. This protein has been demonstrated to specifically associate with the origin of replication of Epstein-Barr virus in human cells, and is thought to be required for DNA replication from viral origin of replication. [provided by RefSeq]
Sequence: MSKPELKEDKMLEVHFVGDDDVLNHILDREGGAKLKKERAQLLVNPKKIIKKPEYDLEEDDQEVLKDQNYVEIMGRDVQESLKNGSATGGGNKVYSFQNRKHSEKMAKLASELAKTPQKSVSFSLKNDPEITINVPQSSKGHSASDKVQPKNNDKSEFLSTAPRSLRKRLIVPRSHSDSESEYSASNSEDDEGVAQEHEEDTNAVIFSQKIQAQNRVVSAPVGKETPSKRMKRDKTSDLVEEYFEAHSSSKVLTSDRTLQKLKRAKLDQQTLRNLLSKVSPSFSAELKQLNQQYEKLFHKWMLQLHLGFNIVLYGLGSKRDLLERFRTTMLQDSIHVVINGFFPGISVKSVLNSITEEVLDHMGTFRSILDQLDWIVNKFKEDSSLELFLLIHNLDSQMLRGEKSQQIIGQLSSLHNIYLIASIDHLNAPLMWDHAKQSLFNWLWYETTTYSPYTEETSYENSLLVKQSGSLPLSSLTHVLRSLTPNARGIFRLLIKYQLDNQDNPSYIGLSFQDFYQQCREAFLVNSDLTLRAQLTEFRDHKLIRTKKGTDGVEYLLIPVDNGTLTDFLEKEEEEAEspecificaciones
AAH14834 | |
4999 | |
Glutathione Sepharose 4 Fast Flow | |
RUO | |
ORC2L | |
Wheat Germ (in vitro) | |
wheat germ expression system |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
89.21kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
ORC2 | |
ORC2L | |
Recombinant | |
Liquid |