missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human NOD2 Synthetic Peptide
Synthetic Peptide
Marca: Invitrogen™ PEP0307
Detalles adicionales : Peso : 0.01000kg
Descripción
NOD2 Synthetic Peptide, PEP-0307, from Invitrogen.
This peptide corresponds to 14 amino acids near the carboxy terminus of human NOD2.PEP-0307 can be used as a blocking peptide with polyclonal antibody PA5-19992.Especificaciones
64127 | |
purified | |
Human NOD2 | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
No | |
Peptide | |
RUO - research use only |
Human | |
Q9HC29 | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
NOD2 | |
0.02% sodium azide | |
50 μg | |
PBS |