Learn More
Novus Biologicals™ Human MMP-17 ELISA Kit (Colorimetric)
Colorimetric ELISA Kit
Marca: Novus Biologicals™ NBP3-06888
Detalles adicionales : Peso : 0.00970kg
Especificaciones
0.16 to 10 ng/mL | |
ELISA Kit (Colorimetric) | |
This kit recognizes Human MMP-17 in samples. No significant cross-reactivity or interference between Human MMP-17 and analogues was observed. | |
4326 | |
1 Kit | |
Storage of components varies. See protocol for specific instructions. | |
Human |
0.09 ng/mL | |
Cell Culture Supernatant, Plasma, Serum | |
ELISA, Sandwich ELISA | |
MMP17 | |
Angiogenesis, Cancer, Cardiovascular Biology, Cell Biology, Cytoskeleton Markers, Signal Transduction | |
matrix metallopeptidase 17 (membrane-inserted), matrix metalloproteinase 17 (membrane-inserted), matrix metalloproteinase-17, membrane-type matrix metalloproteinase 4, membrane-type-4 matrix metalloproteinase, MMP-17, MT4MMP, MT4-MMP, MT-MMP 4, MTMMP4 |