Learn More
Abnova™ Human MKRN2 Full-length ORF (NP_054879.3, 1 a.a. - 416 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00023609-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Members of the makorin family, including MKRN2, have a characteristic zinc finger composition that suggests that they are ribonucleoproteins (Gray et al., 2001 [PubMed 11597136]).[supplied by OMIM]
Sequence: MSTKQITCRYFMHGVCREGSQCLFSHDLANSKPSTICKYYQKGYCAYGTRCRYDHTRPSAAAGGAVGTMAHSVPSPAFHSPHPPSEVTASIVKTNSHEPGKREKRTLVLRDRNLSGMAERKTQPSMVSNPGSCSDPQPSPEMKPHSYLDAIRSGLDDVEASSSYSNEQQLCPYAAAGECRFGDACVYLHGEVCEICRLQVLHPFDPEQRKAHEKICMLTFEHEMEKAFAFQASQDKVCSICMEVILEKASASERRFGILSNCNHTYCLSCIRQWRCAKQFENPIIKSCPECRVISEFVIPSVYWVEDQNKKNELIEAFKQGMGKKACKYFEQGKGTCPFGSKCLYRHAYPDGRLAEPEKPRKQLSSQGTVRFFNSVRLWDFIENRESRHVPNNEDVDMTELGDLFMHLSGVESSEPEspecificaciones
NP_054879.3 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
73.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HSPC070/RNF62 | |
MKRN2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
23609 | |
MKRN2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSTKQITCRYFMHGVCREGSQCLFSHDLANSKPSTICKYYQKGYCAYGTRCRYDHTRPSAAAGGAVGTMAHSVPSPAFHSPHPPSEVTASIVKTNSHEPGKREKRTLVLRDRNLSGMAERKTQPSMVSNPGSCSDPQPSPEMKPHSYLDAIRSGLDDVEASSSYSNEQQLCPYAAAGECRFGDACVYLHGEVCEICRLQVLHPFDPEQRKAHEKICMLTFEHEMEKAFAFQASQDKVCSICMEVILEKASASERRFGILSNCNHTYCLSCIRQWRCAKQFENPIIKSCPECRVISEFVIPSVYWVEDQNKKNELIEAFKQGMGKKACKYFEQGKGTCPFGSKCLYRHAYPDGRLAEPEKPRKQLSSQGTVRFFNSVRLWDFIENRESRHVPNNEDVDMTELGDLFMHLSGVESSEP | |
RUO | |
MKRN2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |