missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human LZTS1 Partial ORF (NP_066300, 514 a.a. - 596 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Especificaciones
Número de acceso | NP_066300 |
---|---|
Para utilizar con (aplicación) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulación | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
ID de gen (Entrez) | 11178 |
Peso molecular | 34.87kDa |
Descripción
Sequence: ERQGHDQMSSGFQHERLVWKEEKEKVIQYQKQLQQSYVAMYQRNQRLEKALQQLARGDSAGEPLEVDLEGADIPYEDIIATEIEspecificaciones
NP_066300 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.87kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
F37/FEZ1 | |
LZTS1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
11178 | |
LZTS1 (Human) Recombinant Protein (Q01) | |
ERQGHDQMSSGFQHERLVWKEEKEKVIQYQKQLQQSYVAMYQRNQRLEKALQQLARGDSAGEPLEVDLEGADIPYEDIIATEI | |
RUO | |
LZTS1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |