missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Livin Synthetic Peptide
Synthetic Peptide
Marca: Invitrogen™ PEP0114
Detalles adicionales : Peso : 0.01000kg
Descripción
Livin Synthetic Peptide, PEP-0114, from Invitrogen.
This peptide corresponds to 35 amino acids of human Livin.PEP-0114 can be used as a blocking peptide with polyclonal antibody PA5-19990.Especificaciones
79444 | |
purified | |
Human Livin | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
No | |
Peptide | |
RUO - research use only |
Human | |
Q96CA5 | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
BIRC7 | |
0.02% sodium azide | |
50 μg | |
PBS |