Learn More
Invitrogen™ Human KCNMA1 (aa 887-902) Synthetic Peptide
Synthetic Peptide
Marca: Invitrogen™ PEP122
Detalles adicionales : CAS : 69-74-9 Peso : 0.01000kg
Descripción
Human Maxi K+ alpha (residues 887-902) Synthetic Peptide, PEP-122, from Invitrogen.
PEP-122 is a 16 amino acid synthetic peptide whose sequences are derived from amino acids 887-902 of human Maxi K+ alpha protein.This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and human NFAT3, catalog # PA1-021. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.Especificaciones
3778 | |
purified | |
Human KCNMA1 (aa 887-902) | |
-20°C. Avoid freeze-thaw cycles. | |
Lyophilized | |
Control | |
Yes | |
Peptide | |
RUO - research use only |
Human | |
Q12791 | |
0.5 mg/mL | |
Synthetic Peptide | |
peptide with no preservative | |
KCNMA1 | |
No preservative | |
50 μg | |
Derived from Amino Acids 887-902 of Human Maxi K+ a Protein |