Learn More
STEMCELL Technologies Human IL-17A ELISA Kit
For detection and measurement of human interleukin 17A
Marca:  STEMCELL Technologies 02036
Detalles adicionales : Peso : 0.03670kg
 Ver más versiones de este producto
Código de producto. 17198041
  • 1231.20€ / paquete de 2
Fecha estimada de envío: 10-05-2024
para ver el stock
Añadir a la cesta
Descripción
Descripción
  • The Human Interleukin 17A (IL-17A) ELISA Kit is designed for the quantitative detection and measurement of human IL-17A in biological fluids such as serum, plasma, and cell culture supernatants
  • IL-17A is secreted by Th17 cells, CD8+ T cells, γδ T cells, natural killer T cells, B cells, innate lymphoid cells, and mesenchymal stromal cells
  • It is a pro-inflammatory cytokine that plays a role in the defense against pathogens, activates the NF-kB, MAPK, and C/EBP pathway
  • The assay is based on the sandwich ELISA method, in which samples are added to ELISA strip plates pre-coated with capture antibodies specific for the cytokine
  • The captured cytokine is detected by addition of a biotinylated detection antibody, followed by streptavidin-horseradish peroxidase (SA-HRP), which binds the biotinylated antibody
  • Addition of the chromogenic enzyme substrate 3,3',5,5' tetramethylbenzidine (TMB) results in a colored product with an intensity directly proportional to the concentration of cytokine in the sample
Especificaciones
Especificaciones
B Cells; Lymphocytes; Mesenchymal Stem and Progenitor Cells; T Cells; T Cells, CD4+; T Cells, CD8+; T Cells, Other Subsets
2 Plates