Learn More
Abnova™ Human H1FOO Full-length ORF (AAH47943.1, 1 a.a. - 207 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00132243-P01.10ug
Description
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Nucleosomes consist of approximately 146 bp of DNA wrapped around a histone octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The chromatin fiber is further compacted through the interaction of a linker histone, H1, with the DNA between the nucleosomes to form higher order chromatin structures. The protein encoded is a member of the histone H1 family. This gene contains introns, unlike most histone genes. The protein encoded is a member of the histone H1 family. The related mouse gene is expressed only in oocytes. [provided by RefSeq]
Sequence: MAPATAPRRAGEAKGKGPKKPSEAKEDPPNVGKVKKAAKRPAKVQKPPPKPGAATEKARKQGGAAKDTRAQSGEARKVPPKPDKAMRAPSSAGGLSRKAKAKGSRSSQGDAEAYRKTKAESKSSKPTASKVKNGAASPTKKKVVAKAKAPKAGQGPNTKAAAPAKGSGSKVVPAHLSRKTEAPKGPRKAGLPIKASSSKVSSQRAEASpecifications
AAH47943.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
47.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC50807/osH1 | |
H1FOO | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
132243 | |
H1FOO (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAPATAPRRAGEAKGKGPKKPSEAKEDPPNVGKVKKAAKRPAKVQKPPPKPGAATEKARKQGGAAKDTRAQSGEARKVPPKPDKAMRAPSSAGGLSRKAKAKGSRSSQGDAEAYRKTKAESKSSKPTASKVKNGAASPTKKKVVAKAKAPKAGQGPNTKAAAPAKGSGSKVVPAHLSRKTEAPKGPRKAGLPIKASSSKVSSQRAEA | |
RUO | |
H1FOO | |
Wheat Germ (in vitro) | |
GST | |
Liquid |