Learn More
Abnova™ Human GGTLA1 Partial ORF (NP_004112.1, 510 a.a. - 586 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00002687-Q01.10ug
Weitere Details : Peso : 0.00010kg
Beschreibung
This gene is a member of a gene family that encodes gamma-glutamyl transpeptidase enzymes. This enzyme consists of a heavy and a light chain, and is able to hydrolyze the gamma-glutamyl moiety of glutathione. It converts leukotriene C4 to leukotriene D4, however, it doesn't convert synthetic substrates that are commonly used to assay gamma-glutamyl transpeptidase. Three transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: GFDLRAAIAAPILHVNSKGCVEYEPNFSQEVQRGLQDRGQNQTQRPFFLNVVQAVSQEGACVYAVSDLRKSGEAAGYSpezifikation
NP_004112.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.21kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
GFDLRAAIAAPILHVNSKGCVEYEPNFSQEVQRGLQDRGQNQTQRPFFLNVVQAVSQEGACVYAVSDLRKSGEAAGY | |
RUO | |
GGT5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
2687 | |
GGTLA1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp566O011/FLJ92733/GGT-REL/GGTLA1 | |
GGT5 | |
Recombinant | |
wheat germ expression system |