Learn More
Abnova™ Human EYA4 Full-length ORF (NP_742101.2, 1 a.a. - 616 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00002070-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a member of the eyes absent (EYA) family of proteins. The encoded protein may act as a transcriptional activator through its protein phosphatase activity, and it may be important for eye development, and for continued function of the mature organ of Corti. Mutations in this gene are associated with postlingual, progressive, autosomal dominant hearing loss at the deafness, autosomal dominant nonsyndromic sensorineural 10 locus. Defects in this gene are also associated with dilated cardiomyopathy 1J. Three transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq]
Sequence: MEDSQDLNEQSVKKTCTESDVSQSQNSRSMEMQDLASPHTLVGGGDTPGSSKLEKSNLSSTSVTTNGTGVSLLAVKTEPLNSSETTATTGDGALDTFTGSVITSSGYSPRSAHQYSPQLYPSKPYPHILSTPAAQTMSAYAGQTQYSGMQQPAVYTAYSQTGQPYSLPTYDLGVMLPAIKTESGLSQTQSPLQSGCLSYSPGFSTPQPGQTPYSYQMPGSSFAPSSTIYANNSVSNSTNFSGSQQDYPSYTAFGQNQYAQYYSASTYGAYMTSNNTADGTPSSTSTYQLQESLPGLTNQPGEFDTMQSPSTPIKDLDERTCRSSGSKSRGRGRKNNPSPPPDSDLERVFVWDLDETIIVFHSLLTGSYAQKYGKDPPMAVTLGLRMEEMIFNLADTHLFFNDLEECDQVHIDDVSSDDNGQDLSTYSFATDGFHAAASSANLCLPTGVRGGVDWMRKLAFRYRRVKELYNTYKNNVGGLLGPAKRDAWLQLRAEIEGLTDSWLTNALKSLSIISTRSNCINVLVTTTQLIPALAKVLLYSLGGAFPIENIYSATKIGKESCFERIMQRFGRKVVYVVIGDGVEEEQAAKKHNMPFWRISSHSDLLALHQALELEYLEspecificaciones
NP_742101.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
93.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CMD1J/DFNA10 | |
EYA4 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
2070 | |
EYA4 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MEDSQDLNEQSVKKTCTESDVSQSQNSRSMEMQDLASPHTLVGGGDTPGSSKLEKSNLSSTSVTTNGTGVSLLAVKTEPLNSSETTATTGDGALDTFTGSVITSSGYSPRSAHQYSPQLYPSKPYPHILSTPAAQTMSAYAGQTQYSGMQQPAVYTAYSQTGQPYSLPTYDLGVMLPAIKTESGLSQTQSPLQSGCLSYSPGFSTPQPGQTPYSYQMPGSSFAPSSTIYANNSVSNSTNFSGSQQDYPSYTAFGQNQYAQYYSASTYGAYMTSNNTADGTPSSTSTYQLQESLPGLTNQPGEFDTMQSPSTPIKDLDERTCRSSGSKSRGRGRKNNPSPPPDSDLERVFVWDLDETIIVFHSLLTGSYAQKYGKDPPMAVTLGLRMEEMIFNLADTHLFFNDLEECDQVHIDDVSSDDNGQDLSTYSFATDGFHAAASSANLCLPTGVRGGVDWMRKLAFRYRRVKELYNTYKNNVGGLLGPAKRDAWLQLRAEIEGLTDSWLTNALKSLSIISTRSNCINVLVTTTQLIPALAKVLLYSLGGAFPIENIYSATKIGKESCFERIMQRFGRKVVYVVIGDGVEEEQAAKKHNMPFWRISSHSDLLALHQALELEYL | |
RUO | |
EYA4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |