missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Exostosin-like 3/EXTL3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
191.00€ - 377.00€
Especificaciones
| Antígeno | Exostosin-like 3/EXTL3 |
|---|---|
| Clon | 311007 |
| Concentración | LYOPH |
| Dilución | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
| Aplicaciones | Western Blot, Immunoprecipitation |
| Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
|---|---|---|---|---|---|---|---|---|---|
| Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
18780117
![]() |
R&D Systems
MAB2635-SP |
25 μg |
191.00€
25 microgramos |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
18182551
![]() |
R&D Systems
MAB2635 |
100 μg |
377.00€
100 microgramos |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
Descripción
Exostosin-like 3/EXTL3 Monoclonal specifically detects Exostosin-like 3/EXTL3 in Human samples. It is validated for Western Blot, Immunoprecipitation.Especificaciones
| Exostosin-like 3/EXTL3 | |
| LYOPH | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Human | |
| O43909 | |
| 2137 | |
| Mouse myeloma cell line NS0-derived recombinant human Exostosin-like 3/EXTL3 Thr52-Ile919 Accession # O43909 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Exostosin-like 3/EXTL3 in direct ELISAs and Western blots. |
| 311007 | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| botv, DKFZp686C2342, EC 2.4.1.223, exostoses (multiple)-like 3, Exostosin like 3, exostosin-like 3, EXTL1L, EXTL3, EXTR1BOTV, EXT-related protein 1, Glucuronyl-galactosyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase, Hereditary multiple exostoses gene isolog, KIAA0519, Multiple exostosis-like protein 3, Putative tumor suppressor protein EXTL3, REG, reg receptor, REGR, RPR | |
| EXTL3 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title