missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Exostosin-like 3/EXTL3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marca: R&D Systems MAB2635
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Exostosin-like 3/EXTL3 Monoclonal specifically detects Exostosin-like 3/EXTL3 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Especificaciones
| Exostosin-like 3/EXTL3 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| O43909 | |
| EXTL3 | |
| Mouse myeloma cell line NS0-derived recombinant human Exostosin-like 3/EXTL3 Thr52-Ile919 Accession # O43909 | |
| 100 μg | |
| Primary | |
| Detects human Exostosin-like 3/EXTL3 in direct ELISAs and Western blots. | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| 311007 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| botv, DKFZp686C2342, EC 2.4.1.223, exostoses (multiple)-like 3, Exostosin like 3, exostosin-like 3, EXTL1L, EXTL3, EXTR1BOTV, EXT-related protein 1, Glucuronyl-galactosyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase, Hereditary multiple exostoses gene isolog, KIAA0519, Multiple exostosis-like protein 3, Putative tumor suppressor protein EXTL3, REG, reg receptor, REGR, RPR | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2137 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido