missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CHML Partial ORF (NP_001812, 68 a.a. - 177 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00001122-Q01.25ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The product of the CHML gene supports geranylgeranylation of most Rab proteins and may substitute for REP-1 in tissues other than retina. CHML is localized close to the gene for Usher syndrome type II. [provided by RefSeq]
Sequence: IGEESTVVWQDLIHETEEAITLRKKDETIQHTEAFCYASQDMEDNVEEIGALQKNPSLGVSNTFTEVLDSALPEESQLSYFNSDEMPAKHTQKSDTEISLEVTDVEESVEEspecificaciones
NP_001812 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
IGEESTVVWQDLIHETEEAITLRKKDETIQHTEAFCYASQDMEDNVEEIGALQKNPSLGVSNTFTEVLDSALPEESQLSYFNSDEMPAKHTQKSDTEISLEVTDVEESVE | |
RUO | |
CHML | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
1122 | |
CHML (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ10071/FLJ13361/REP2 | |
CHML | |
Recombinant | |
wheat germ expression system |