missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CDK10 Full-length ORF (NP_003665.2, 1 a.a. - 331 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00008558-P01.10ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The protein encoded by this gene belongs to the CDK subfamily of the Ser/Thr protein kinase family. The CDK subfamily members are highly similar to the gene products of S. cerevisiae cdc28, and S. pombe cdc2, and are known to be essential for cell cycle progression. This kinase has been shown to play a role in cellular proliferation and its function is limited to cell cycle G2-M phase. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: MGRCRSVKEFEKLNRIGEGTYGIVYRARDTQTDEIVALKKVRMDKEKDGIPISSLREITLLLRLRHPNIVELKEVVVGNHLESIFLVMGYCEQDLASLLENMPTPFSEAQVKCIVLQVLRGLQYLHRNFIIHRDLKVSNLLMTDKGCVKTADFGLARAYGVPVKPMTPKVVTLWYRAPELLLGTTTQTTSIDMWAVGCILAELLAHRPLLPGTSEIHQIDLIVQLLGTPSENIWPGFSKLPLVGQYSLRKQPYNNLKHKFPWLSEAGLRLLHFLFMYDPKKRATAGDCLESSYFKEKPLPCEPELMPTFPHHRNKRAAPATSEGQSKRCKPEspecificaciones
NP_003665.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
64kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PISSLRE | |
CDK10 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
8558 | |
CDK10 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MGRCRSVKEFEKLNRIGEGTYGIVYRARDTQTDEIVALKKVRMDKEKDGIPISSLREITLLLRLRHPNIVELKEVVVGNHLESIFLVMGYCEQDLASLLENMPTPFSEAQVKCIVLQVLRGLQYLHRNFIIHRDLKVSNLLMTDKGCVKTADFGLARAYGVPVKPMTPKVVTLWYRAPELLLGTTTQTTSIDMWAVGCILAELLAHRPLLPGTSEIHQIDLIVQLLGTPSENIWPGFSKLPLVGQYSLRKQPYNNLKHKFPWLSEAGLRLLHFLFMYDPKKRATAGDCLESSYFKEKPLPCEPELMPTFPHHRNKRAAPATSEGQSKRCKP | |
RUO | |
CDK10 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |