Learn More
Abnova™ Human CDH8 Full-length ORF (NP_001787.2, 1 a.a. - 799 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00001006-P01.25ug
Descripción
This gene encodes a type II classical cadherin from the cadherin superfamily, integral membrane proteins that mediate calcium-dependent cell-cell adhesion. Mature cadherin proteins are composed of a large N-terminal extracellular domain, a single membrane-spanning domain, and a small, highly conserved C-terminal cytoplasmic domain. The extracellular domain consists of 5 subdomains, each containing a cadherin motif, and appears to determine the specificity of the protein's homophilic cell adhesion activity. Type II (atypical) cadherins are defined based on their lack of a HAV cell adhesion recognition sequence specific to type I cadherins. This particular cadherin is expressed in brain and is putatively involved in synaptic adhesion, axon outgrowth and guidance. [provided by RefSeq]
Sequence: MPERLAEMLLDLWTPLIILWITLPPCIYMAPMNQSQVLMSGSPLELNSLGEEQRILNRSKRGWVWNQMFVLEEFSGPEPILVGRLHTDLDPGSKKIKYILSGDGAGTIFQINDVTGDIHAIKRLDREEKAEYTLTAQAVDWETSKPLEPPSEFIIKVQDINDNAPEFLNGPYHATVPEMSILGTSVTNVTATDADDPVYGNSAKLVYSILEGQPYFSIEPETAIIKTALPNMDREAKEEYLVVIQAKDMGGHSGGLSGTTTLTVTLTDVNDNPPKFAQSLYHFSVPEDVVLGTAIGRVKANDQDIGENAQSSYDIIDGDGTALFEITSDAQAQDGIIRLRKPLDFETKKSYTLKVEAANVHIDPRFSGRGPFKDTATVKIVVEDADEPPVFSSPTYLLEVHENAALNSVIGQVTARDPDITSSPIRFSIDRHTDLERQFNINADDGKITLATPLDRELSVWHNITIIATEIRNHSQISRVPVAIKVLDVNDNAPEFASEYEAFLCENGKPGQVIQTVSAMDKDDPKNGHYFLYSLLPEMVNNPNFTIKKNEDNSLSILAKHNGFNRQKQEVYLLPIIISDSGNPPLSSTSTLTIRVCGCSNDGVVQSCNVEAYVLPIGLSMGALIAILACIILLLVIVVLFVTLRRHKNEPLIIKDDEDVRENIIRYDDEGGGEEDTEAFDIATLQNPDGINGFLPRKDIKPDLQFMPRQGLAPVPNGVDVDEFINVRLHEADNDPTAPPYDSIQIYGYEGRGSVAGSLSSLESTTSDSDQNFDYLSDWGPRFKRLGELYSVGESDKETEspecificaciones
NP_001787.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
114.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
Nbla04261 | |
CDH8 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
1006 | |
CDH8 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MPERLAEMLLDLWTPLIILWITLPPCIYMAPMNQSQVLMSGSPLELNSLGEEQRILNRSKRGWVWNQMFVLEEFSGPEPILVGRLHTDLDPGSKKIKYILSGDGAGTIFQINDVTGDIHAIKRLDREEKAEYTLTAQAVDWETSKPLEPPSEFIIKVQDINDNAPEFLNGPYHATVPEMSILGTSVTNVTATDADDPVYGNSAKLVYSILEGQPYFSIEPETAIIKTALPNMDREAKEEYLVVIQAKDMGGHSGGLSGTTTLTVTLTDVNDNPPKFAQSLYHFSVPEDVVLGTAIGRVKANDQDIGENAQSSYDIIDGDGTALFEITSDAQAQDGIIRLRKPLDFETKKSYTLKVEAANVHIDPRFSGRGPFKDTATVKIVVEDADEPPVFSSPTYLLEVHENAALNSVIGQVTARDPDITSSPIRFSIDRHTDLERQFNINADDGKITLATPLDRELSVWHNITIIATEIRNHSQISRVPVAIKVLDVNDNAPEFASEYEAFLCENGKPGQVIQTVSAMDKDDPKNGHYFLYSLLPEMVNNPNFTIKKNEDNSLSILAKHNGFNRQKQEVYLLPIIISDSGNPPLSSTSTLTIRVCGCSNDGVVQSCNVEAYVLPIGLSMGALIAILACIILLLVIVVLFVTLRRHKNEPLIIKDDEDVRENIIRYDDEGGGEEDTEAFDIATLQNPDGINGFLPRKDIKPDLQFMPRQGLAPVPNGVDVDEFINVRLHEADNDPTAPPYDSIQIYGYEGRGSVAGSLSSLESTTSDSDQNFDYLSDWGPRFKRLGELYSVGESDKET | |
RUO | |
CDH8 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |