Learn More
Abnova™ Human CAPZA3 Full-length ORF (AAH16745.1, 1 a.a. - 299 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00093661-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes an actin capping protein, one of the F-actin capping protein alpha subunit family. The encoded protein is predominantly localized to the neck region of ejaculated sperm, other immunohistochemical signals were found in the tail and postacrosomal regions. The encoded protein may also form heterodimers of alpha and beta subunits. This protein may be important in determining sperm architecture and male fertility. [provided by RefSeq]
Sequence: MTLSVLSRKDKERVIRRLLLQAPPGEFVNAFDDLCLLIRDEKLMHHQGECAGHQHCQKYSVPLCIDGNPVLLSHHNVMGDYRFFDHQSKLSFKYYLLQNQLKDIQSHGIIQNEAEYLRVVLLCALKLYVNDHYPKGNCNMLRKTVKSKEYLIACIEDHNYETGECWNGLWKSKWIFQVNPFLTQVTGRIFVQAHFFRCVNLHIEISKDLKESLEIVNQAQLALSFARLVEEQENKFQAAVLEELQELSNEALRKILRRDLPVTRTLIDWHRILSDLNLVMYPKLGYVIYSRSVLCNWIIEspecificaciones
AAH16745.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
61.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CAPPA3/Gsg3 | |
CAPZA3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
93661 | |
CAPZA3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MTLSVLSRKDKERVIRRLLLQAPPGEFVNAFDDLCLLIRDEKLMHHQGECAGHQHCQKYSVPLCIDGNPVLLSHHNVMGDYRFFDHQSKLSFKYYLLQNQLKDIQSHGIIQNEAEYLRVVLLCALKLYVNDHYPKGNCNMLRKTVKSKEYLIACIEDHNYETGECWNGLWKSKWIFQVNPFLTQVTGRIFVQAHFFRCVNLHIEISKDLKESLEIVNQAQLALSFARLVEEQENKFQAAVLEELQELSNEALRKILRRDLPVTRTLIDWHRILSDLNLVMYPKLGYVIYSRSVLCNWII | |
RUO | |
CAPZA3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |