Learn More
Abnova™ Human CACNA1E Partial ORF (NP_000712, 1901 a.a. - 1999 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00000777-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes an alpha-1 subunit of a voltage-dependent calcium channel. Calcium channels mediate the influx of calcium ions into the cell upon membrane polarization. The alpha-1 subunit consists of 24 transmembrane segments and forms the pore through which ions pass into the cell. The calcium channel consists of a complex of alpha-1, alpha-2/delta, beta, and gamma subunits in a 1:1:1:1 ratio. There are multiple isoforms of each of these proteins, either encoded by different genes or the result of alternative splicing of transcripts. Alternate transcriptional splice variants of the gene described here have been observed but have not been thoroughly characterized. [provided by RefSeq]
Sequence: RMEPSSLPQEIIANAKALPYLQQDPVSGLSGRSGYPSMSPLSPQDIFQLACMDPADDGQFQERQSLVVTDPSSMRRSFSTIRDKRSNSSWLEEFSMERSEspecificaciones
NP_000712 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RMEPSSLPQEIIANAKALPYLQQDPVSGLSGRSGYPSMSPLSPQDIFQLACMDPADDGQFQERQSLVVTDPSSMRRSFSTIRDKRSNSSWLEEFSMERS | |
RUO | |
CACNA1E | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
777 | |
CACNA1E (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BII/CACH6/CACNL1A6/Cav2.3 | |
CACNA1E | |
Recombinant | |
wheat germ expression system |