Learn More
Abnova™ Human C19orf2 Partial ORF (NP_003787, 371 a.a. - 475 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00008725-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene binds to RNA polymerase II subunit 5 (RPB5) and negatively modulates transcription through its binding to RPB5. The encoded protein seems to have inhibitory effects on various types of activated transcription, but it requires the RPB5-binding region. This protein acts as a corepressor. It is suggested that it may require signaling processes for its function or that it negatively modulates genes in the chromatin structure. Two alternatively spliced transcript variants encoding different isoforms have been described for this gene. [provided by RefSeq]
Sequence: NSTGSGHSAQELPTIRTPADIYRAFVDVVNGEYVPRKSILKSRSRENSVCSDTSESSAAEFDDRRGVLRSISCEEATCSDTSESILEEEPQENQKKLLPLSVTPEEspecificaciones
NP_003787 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.29kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
NSTGSGHSAQELPTIRTPADIYRAFVDVVNGEYVPRKSILKSRSRENSVCSDTSESSAAEFDDRRGVLRSISCEEATCSDTSESILEEEPQENQKKLLPLSVTPE | |
RUO | |
C19orf2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
8725 | |
C19orf2 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ10575/NNX3/RMP/URI | |
C19orf2 | |
Recombinant | |
wheat germ expression system |