Learn More
Abnova™ Human AURKC Partial ORF (NP_003151, 186 a.a. - 275 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00006795-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a member of the Aurora subfamily of serine/threonine protein kinases. The encoded protein is a chromosomal passenger protein that forms complexes with Aurora-B and inner centromere proteins and may play a role in organizing microtubules in relation to centrosome/spindle function during mitosis. This gene is overexpressed in several cancer cell lines, suggesting an involvement in oncogenic signal transduction. Alternative splicing results in multiple transcript variants. [provided by RefSeq]
Sequence: VDLWCIGVLCYELLVGYPPFESASHSETYRRILKVDVRFPLSMPLGARDLISRLLRYQPLERLPLAQILKHPWVQAHSRRVLPPCAQMASEspecificaciones
NP_003151 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.53kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VDLWCIGVLCYELLVGYPPFESASHSETYRRILKVDVRFPLSMPLGARDLISRLLRYQPLERLPLAQILKHPWVQAHSRRVLPPCAQMAS | |
RUO | |
AURKC | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
6795 | |
AURKC (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AIE2/AIK3/AurC/STK13/aurora-C | |
AURKC | |
Recombinant | |
wheat germ expression system |