Learn More
Abnova™ Human APOA4 Full-length ORF (AAH74764.1, 1 a.a. - 396 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00000337-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Apoliprotein (apo) A-IV gene contains 3 exons separated by two introns. A sequence polymorphism has been identified in the 3'UTR of the third exon. The primary translation product is a 396-residue preprotein which after proteolytic processing is secreted its primary site of synthesis, the intestine, in association with chylomicron particles. Although its precise function is not known, apo A-IV is a potent activator of lecithin-cholesterol acyltransferase in vitro. [provided by RefSeq]
Sequence: MFLKAVVLTLALVAVAGARAEVSADQVATVMWDYFSQLSNNAKEAVEHLQKSELTQQLNALFQDKLGEVNTYAGDLQKKLVPFATELHERLAKDSEKLKEEIGKELEELRARLLPHANEVSQKIGDNLRELQQRLEPYADQLRTQVNTQAEQLRRQLTPYAQRMERVLRENADSLQASLRPHADELKAKIDQNVEELKGRLTPYADEFKVKIDQTVEELRRSLAPYAQDTQEKLNHQLEGLTFQMKKNAEELKARISASAEELRQRLAPLAEDVRGNLRGNTEGLQKSLAELGGHLDQQVEEFRRRVEPYGENFNKALVQQMEQLRQKLGPHAGDVEGHLSFLEKDLRDKVNSFFSTFKEKESQDKTLSLPELEQQQEQQQEQQQEQVQMLAPLESEspecificaciones
AAH74764.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
71.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
RUO | |
APOA4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
337 | |
APOA4 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC142154/MGC142156 | |
APOA4 | |
Recombinant | |
wheat germ expression system |