Learn More
ACROBiosystems Human Angiopoietin-like 4 / ANGPTL4 (166-406) Protein, His Tag
Marca: ACROBiosystems AN4-H5243-100UG
Detalles adicionales : Peso : 0.00001kg
Descripción
Angiopoietin-related protein 4 (ANGPTL4) is also known as Hepatic fibrinogen / angiopoietin - related protein (HFARP), ARP4, PGAR. ANGPTL4 contains one fibrinogen C-terminal domain. ANGPTL4 is a homooligomer , and the homooligomer undergoes proteolytic processing to release its carboxyl fibrinogen-like domain, which circulates as a monomer. The homooligomer unprocessed form is able to interact with the extracellular matrix. ANGPTL4 may act as a regulator of angiogenesis and modulate tumorigenesis. ANGPTL4 inhibits proliferation, migration, and tubule formation of endothelial cells and reduces vascular leakage. ANGPTL4 may exert a protective function on endothelial cells through an endocrine action.
This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 27.9 kDa. The protein migrates as 33-38 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
NP_647475.1 | |
27.9 kDa | |
100 µg | |
Pro 166 - Ser 406 | |
ANGPTL4,ANGPTL2,ARP4,FIAF,HFARP,NL2,PGAR,pp1158,Angiopoietin like 4 | |
Unconjugated | |
50 mM MOPS, 100 mM NaCl, pH7.2 | |
HEK293 | |
Human |
Powder | |
>95% as determined by SDS-PAGE | |
Human Angiopoietin-like 4 (166-406), His Tag (AN4-H5243) is expressed from human 293 cells (HEK293). It contains AA Pro 166 - Ser 406 (Accession # Q9BY76-1). | |
-20°C | |
Angiopoietin-like 4 | |
His Tag | |
Less than 1.0 EU / µg by the LAL method. | |
Bio-Markers and CD Antigens |