Learn More
Invitrogen™ Human Aggrecan (aa 89-106) Synthetic Peptide
Synthetic Peptide
Marca: Invitrogen™ PEP236
Detalles adicionales : CAS : 69-74-9 Peso : 0.01000kg
Descripción
Human Aggrecan (residues 89-106) Synthetic Peptide, PEP-236, from Invitrogen.
PEP-236 is a 19 amino acid synthetic peptide corresponding to amino acid residues 89-106 from human aggrecan. The sequence of this peptide is (amino to carboxy terminus): C A (89) T E G Q V R V N S I Y Q D K V S L (106).This peptide may be used for neutralization and control experiments with a polyclonal antibody that reacts with this product. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.Especificaciones
176 | |
purified | |
Human Aggrecan (aa 89-106) | |
-20°C. Avoid freeze-thaw cycles. | |
Lyophilized | |
Control | |
Yes | |
Peptide | |
RUO - research use only |
Human | |
P16112 | |
0.5 mg/mL | |
Synthetic Peptide | |
peptide with no preservative | |
ACAN | |
No preservative | |
50 μg | |
C A(89) T E G Q V R V N S I Y Q D K V S L(106) |