Learn More
ACROBiosystems Human Activin RIIB / ACVR2B Protein, His Tag
Marca: ACROBiosystems ACB-H5226-200UG
Detalles adicionales : Peso : 0.00001kg
Descripción
Activin receptor type-2B (ACVR2B) is also known as ActR-IIB and MGC116908, ACVR2B is an activin type 2 receptor. Activins are dimeric growth and differentiation factors which belong to the transforming growth factor-beta (TGF-beta) superfamily of structurally related signaling proteins. Activins signal through a heteromeric complex of receptor serine kinases which include at least two type I (I and IB) and two type II (II and IIB) receptors. These receptors are all transmembrane proteins, composed of a ligand-binding extracellular domain with cysteine-rich region, a transmembrane domain, and a cytoplasmic domain with predicted serine/threonine specificity. Type I receptors are essential for signaling; and type II receptors are required for binding ligands and for expression of type I receptors. Type I and II receptors form a stable complex after ligand binding, resulting in phosphorylation of type I receptors by type II receptors. Type II receptors are considered to be constitutively active kinases. This gene encodes activin A type IIB receptor, which displays a 3- to 4-fold higher affinity for the ligand than activin A type II receptor. Defects in ACVR2B are the cause of visceral heterotaxy autosomal type 4 (HTX4).
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 14.5 kDa. The protein migrates as 22-44 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
NP_001097.2 | |
14.5 kDa | |
200 µg | |
Ser 19 - Thr 137 | |
ACVR2B,ACTRIIB,MGC116908 | |
Unconjugated | |
PBS, pH7.4 | |
HEK293 | |
Human |
Powder | |
>90% as determined by SDS-PAGE | |
Human Activin RIIB, His Tag (ACB-H5226) is expressed from human 293 cells (HEK293). It contains AA Ser 19 - Thr 137 (Accession # Q13705-1). | |
-20°C | |
Activin RIIB | |
His Tag | |
Less than 1.0 EU / µg by the LAL method. | |
Other Recombinant Proteins |