Learn More
Thermo Scientific™ HTM Broth
Perform high quality susceptibility testing with a range of supplies, including the Thermo Scientific™ HTM Broth, to enhance your Thermo Scientific™ Sensititre™ test program.
67.95€
Especificaciones
Tipo de producto | Caldo HTM |
---|---|
Tipo de prueba | Susceptibilidad microbiana |
N.° por paquete | 10 |
Tipo de empaquetado | Frasco de 11 ml |
Cantidad | 11 mL |
Código de producto | Marca | Descripción | Cantidad | Precio | Cantidad y disponibilidad | ||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Descripción | Cantidad | Precio | Cantidad y disponibilidad | ||||
16531840
|
Thermo Scientific™
YT3470 |
Caldo HTM | 11 mL |
67.95€
paquete de 2 |
Fecha estimada de envío: 17-05-2024
Inicie sesión para ver el stock |
||||
Descripción
Perform high-quality susceptibility testing with a complete range of supplies, including the Thermo Scientific™ HTM Broth, to complement and enhance your Thermo Scientific™ Sensititre™ test program.
Not all products are available for sale in all territories. Please inquire.VersaTREK™ and Sensititre products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Especificaciones
Caldo HTM | |
10 | |
11 mL |
Susceptibilidad microbiana | |
Frasco de 11 ml |