missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP60 Mouse anti-Human, Mouse, Rat, Clone: 2F10E7, Proteintech
Mouse Monoclonal Antibody
140.00€ - 399.00€
Especificaciones
Antígeno | HSP60 |
---|---|
Clon | 2F10E7 |
Concentración | 0.95 mg/mL |
Aplicaciones | Immunocytochemistry, Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation |
Clasificación | Monoclonal |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
16887555
|
Proteintech
66041-1-IG-20UL |
20 μL |
140.00€
20 microlitros |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
16877555
|
Proteintech
66041-1-IG-150UL |
150 μL |
399.00€
150 microlitros |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
Descripción
This gene encodes a member of the chaperonin family. The encoded mitochondrial protein may function as a signaling molecule in the innate immune system. This protein is essential for the folding and assembly of newly imported proteins in the mitochondria. This gene is adjacent to a related family member and the region between the 2 genes functions as a bidirectional promoter. Several pseudogenes have been associated with this gene. Two transcript variants encoding the same protein have been identified for this gene. Mutations associated with this gene cause autosomal recessive spastic paraplegia 13.Especificaciones
HSP60 | |
0.95 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Heat shock protein 60, HSP 60, HSP60, HSP65, HSPD1 | |
HSPD1 | |
IgG1 | |
Protein A | |
Antibody |
2F10E7 | |
Immunocytochemistry, Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Human, Rat, Mouse | |
P10809, P63038, P63039 | |
15510, 3329, 63868 | |
HSP60 Fusion Protein Ag7848 | |
Primary | |
-20°C | |
HSPD1 |