missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP60 Mouse anti-Human, Mouse, Rat, Clone: 2F10E7, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66041-1-IG-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This gene encodes a member of the chaperonin family. The encoded mitochondrial protein may function as a signaling molecule in the innate immune system. This protein is essential for the folding and assembly of newly imported proteins in the mitochondria. This gene is adjacent to a related family member and the region between the 2 genes functions as a bidirectional promoter. Several pseudogenes have been associated with this gene. Two transcript variants encoding the same protein have been identified for this gene. Mutations associated with this gene cause autosomal recessive spastic paraplegia 13.Especificaciones
| HSP60 | |
| Monoclonal | |
| 0.95 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P10809, P63038, P63039 | |
| HSPD1 | |
| HSP60 Fusion Protein Ag7848 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 2F10E7 | |
| Unconjugated | |
| HSPD1 | |
| Heat shock protein 60, HSP 60, HSP60, HSP65, HSPD1 | |
| Mouse | |
| Protein A | |
| RUO | |
| 15510, 3329, 63868 | |
| -20°C | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido