Learn More
HSP47 Mouse, anti-Human, Unconjugated, Clone: 2A8F6, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 67863-1-IG-150UL
Detalles adicionales : Peso : 0.00020kg
Descripción
The HSP47 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human HSP47. This antibody recognizes human antigen. The HSP47 antibody has been validated for the following applications: WB,ELISA analysis.
Especificaciones
HSP47 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, ELISA | |
2A8F6 | |
Unconjugated | |
150 μL | |
871 | |
Liquid |