missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXD8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA535217
Detalles adicionales : Peso : 0.01000kg
Descripción
This gene belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, located on different chromosomes, consisting of 9 to 11 genes arranged in tandem. This gene is one of several homeobox HOXD genes located in a cluster on chromosome 2. Deletions that remove the entire HOXD gene cluster or the 5' end of this cluster have been associated with severe limb and genital abnormalities. In addition to effects during embryogenesis, this particular gene may also play a role in adult urogenital tract function.Especificaciones
HOXD8 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
P13378 | |
HOXD8 | |
KLH conjugated synthetic peptide between 243-270 amino acids from the C-terminal region of human HOXD8 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry, Western Blot | |
Unconjugated | |
HOXD8 | |
4921540P06Rik; AI047735; homeo box 4E; homeo box D8; homeobox D8; homeobox protein 5.4; homeobox protein Hox-4.3; Homeobox protein Hox-4E; homeobox protein Hox-5.4; Homeobox protein Hox-D8; homeobox protein Hox-D8-like; HOX4; Hox-4.3; Hox-4.5; HOX4E; HOX5.4; Hox-5.4; HOXD8; Hoxd-8; LOC100912936 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
3234 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |