Learn More
HOMEZ Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA557609
Detalles adicionales : Peso : 0.02000kg
Descripción
Immunogen sequence: PDIQPLERYW AAHQQLRETD IPQLSQASRL STQQVLDWFD SRLPQPAEVV VCLDE Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 89%.
May function as a transcriptional regulator and a vertebrate homeobox gene.Especificaciones
HOMEZ | |
Polyclonal | |
Unconjugated | |
HOMEZ | |
homeobox and leucine zipper encoding; homeobox and leucine zipper protein Homez; Homeodomain leucine zipper-containing factor; homeodomain leucine zipper-encoding; homeodomain leucine zipper-encoding gene; homeodomain-leucine zipper protein HOMEZ; HOMEZ; KIAA1443; mKIAA1443 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57594 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IX15 | |
HOMEZ | |
Recombinant protein corresponding to Human HOMEZ | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only