missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HO-1/HMOX1 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 2D10A5, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66743-1-IG-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a protein serine/threonine kinase that is activated when bound to the GTP-bound form of Rho. The small GTPase Rho regulates formation of focal adhesions and stress fibers of fibroblasts, as well as adhesion and aggregation of platelets and lymphocytes by shuttling between the inactive GDP-bound form and the active GTP-bound form. Rho is also essential in cytokinesis and plays a role in transcriptional activation by serum response factor. This protein, a downstream effector of Rho, phosphorylates and activates LIM kinase, which in turn, phosphorylates cofilin, inhibiting its actin-depolymerizing activity.Especificaciones
HO-1/HMOX1 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P06762, P09601, P14901, P32394 | |
HMOX1 | |
HO-1/HMOX1 Fusion Protein Ag21296 | |
150 μL | |
Primary | |
Human, Mouse, Porcine, Rat | |
Antibody | |
IgG2a |
Immunohistochemistry (Paraffin), Western Blot | |
2D10A5 | |
Unconjugated | |
HMOX1 | |
bK286B10, heme oxygenase (decycling) 1, Heme oxygenase 1, HMOX1, HO, HO 1, HO1, HSP32 | |
Mouse | |
Protein A | |
RUO | |
15368, 24451, 3162, 445512 | |
-20°C | |
Liquid |