missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA-F Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA543345
Detalles adicionales : Peso : 0.00010kg
Descripción
Peptide sequence: PWVEQEGPQY WEWTTGYAKA NAQTDRVALR NLLRRYNQSE AGSHTLQGMN Sequence homology: Human: 100%; Pig: 100%
HLA-F Polyclonal antibody specifically detects HLA-F in Human samples. It is validated for Western Blot.Especificaciones
HLA-F | |
Polyclonal | |
Unconjugated | |
HLA-F | |
CDA12; HLA class I histocompatibility antigen, alpha chain F; HLA class I molecule; HLA F antigen; HLA-5.4; HLA-CDA12; HLAF; HLA-F; Leukocyte antigen F; major histocompatibility complex, class I, F; MHC class I antigen F; MHC class Ib antigen | |
Rabbit | |
Affinity Chromatography | |
RUO | |
3134 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P30511 | |
HLA-F | |
Synthetic peptide directed towards the N-terminal of human HLA-F | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |