missing translation for 'onlineSavingsMsg'
Learn More
Learn More
hIST1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 19842-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
RAB10 belongs to the RAS superfamily of small GTPases. RAB proteins localize to exocytic and endocytic compartments and regulate intracellular vesicle trafficking.Especificaciones
hIST1 | |
Polyclonal | |
Unconjugated | |
IST1 | |
hIST1, IST1, IST1 homolog, KIAA0174, OLC1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
307833, 71955, 9798 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P53990, Q568Z6, Q9CX00 | |
Ist1 | |
hIST1 Fusion Protein Ag13874 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |