missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Julabo™ Heating Immersion Circulator
Marca: Julabo™ 9116000
Detalles adicionales : Peso : 0.50000kg
Especificaciones
DIN 12876-1 S1 | |
20°C to 100°C | |
ED | |
±0.03°C | |
13 cm | |
3.3 kg | |
230 V |
15L/min. | |
LED | |
100°C | |
15 cm | |
33 cm | |
230 V 50 Hz 9 A, 230 V 60 Hz 8/9 A | |
50/60 Hz |