missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HAUS4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 20104-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a subunit of the centrosome complex termed the human augmin complex. The encoded protein localizes to the spindle microtubules and may play a role in mitotic spindle assembly and maintece of centrosome integrity during cell division. Alternate splicing results in multiple transcript variants. A pseudogene of this gene is found on chromosome 1.Especificaciones
HAUS4 | |
Polyclonal | |
Unconjugated | |
HAUS4 | |
C14orf94, HAUS4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
219072, 305882, 54930 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8BFT2, Q9H6D7 | |
Haus4 | |
HAUS4 Fusion Protein Ag13717 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |