missing translation for 'onlineSavingsMsg'
Learn More
Learn More
H3K4me3 Recombinant Rabbit Monoclonal Antibody (9H28L57), ChIP-Verified, Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marca: Invitrogen 70395420UG
Detalles adicionales : Peso : 0.01000kg
Descripción
This antibody is predicted to react with non-human primate.
Histone H3 is one of the DNA-binding proteins found in the chromatin of all eukaryotic cells. H3 along with four core histone proteins binds to DNA forming the structure of the nucleosome. Histones play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. Post translationally, histones are modified in a variety of ways to either directly change the chromatin structure or allow for the binding of specific transcription factors. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of post-translational modification that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine.Especificaciones
H3K4me3 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P68431, P84243, Q6NXT2, Q71DI3 | |
H3-3A, H3-3B, H3-5, H3C1, H3C10, H3C14, H3C15, H3C2, H3C7 | |
Peptide corresponding to human H3K4me3 [aa2-aa9] | |
20 μg | |
Primary | |
Human | |
Antibody | |
IgG |
ChIP Assay | |
9H28L57 | |
Unconjugated | |
H3-3A | |
B0035.7; BUR5; CELE_T10C6.12; CG31613; CG31613 gene product from transcript CG31613-RA; CG31613-PA; CG33803; CG33806; CG33809; CG33812; CG33815; CG33818; CG33821; CG33824; CG33827; CG33830; CG33833; CG33836; CG33839; CG33842; CG33845; CG33848; CG33851; CG33854; CG33857; CG33860; CG33863; CG33866; Dmel\CG31613; Dmel_CG31613; F07B7.10; F07B7.3; F08G2.2; F17E9.13; F35H10.1; F45F2.4; F54E12.5; F55G1.10; H02I12.7; H3; H3 histone; H3 histone family, member A; H3 histone family, member I; H3 histone family, member K; H3 histone family, member L; H3 histone family, member M; H3 histone, family 2; H3 histone, family 3A; H3 histone, family 3B; H3 histone, family 3B (H3.3B); H3 histone, family 3B.1; H3 histone, family 3C; H3 K4; H3.1-221; H3.1-291; H3.1-I; H3.2; H3.2-221; H3.2-614; H3.2-615; H3.2-616; h3.2a; H3.3A; H3.3B; H3.5; H3/A; H3/b; H3/d; H3/f; H3/i; H3/j; H3/k; H3/l; H3/M; H3/n; H3/o; H3-143; H3-291; H3-3A; H3-3B; H3-53; H3-614; H3a; H3b; H3-B; H3c1; H3c10; H3C11; H3C12; H3C2; H3C3; H3C4; | |
Rabbit | |
Protein A | |
RUO | |
126961, 3020, 3021, 333932, 440093, 8350, 8357, 8358, 8968 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |