missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GYPE Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA5121481
Detalles adicionales : Peso : 0.01000kg
Descripción
GYPE (Glycophorin E (MNS Blood Group)) is a Protein Coding gene. Diseases associated with GYPE include Malaria and Hemidystonia. An important paralog of this gene is GYPA. The protein encoded by this gene is a sialoglycoprotein and a type I membrane protein. It is a member of a gene family with GPA and GPB genes. This encoded protein might carry the M blood group antigen. GYPA, GYPB, and GYPE are organized in tandem on chromosome 4. This gene might have derived from an ancestral gene common to the GPB gene by gene duplication. Two alternatively spliced transcript variants encoding the same protein have been described for this gene.Especificaciones
GYPE | |
Polyclonal | |
Unconjugated | |
GYPE | |
Glycophorin E | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2996 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
P15421 | |
GYPE | |
Ser20-Ile70 (Accession P15421), with N-terminal His and GST Tag | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |