missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GTF2H2C Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17959-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Description
Component of the core-TFIIH basal transcription factor involved in nucleotide excision repair (NER) of DNA and, when complexed to CAK, in RNA transcription by RNA polymerase II.Specifications
GTF2H2C | |
Polyclonal | |
Unconjugated | |
GTF2H2C | |
GTF2H2C, GTF2H2D | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
728340 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6P1K8 | |
GTF2H2C | |
GTF2H2C Fusion Protein Ag12498 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |