missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSTM4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 16766-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Cytosolic and membrane-bound forms of glutathione S-transferase are encoded by two distinct supergene families. At present, eight distinct classes of the soluble cytoplasmic mammalian glutathione S-transferases have been identified: alpha, kappa, mu, omega, pi, sigma, theta and zeta. This gene encodes a glutathione S-transferase that belongs to the mu class. The mu class of enzymes functions in the detoxification of electrophilic compounds, including carcinogens, therapeutic drugs, environmental toxins and products of oxidative stress, by conjugation with glutathione. The genes encoding the mu class of enzymes are organized in a gene cluster on chromosome 1p13.3 and are known to be highly polymorphic. These genetic variations can change an individual′s susceptibility to carcinogens and toxins as well as affect the toxicity and efficacy of certain drugs. Diversification of these genes has occurred in regions encoding substrate-binding domains, as well as in tissue expression patterns, to accommodate an increasing number of foreign compounds. Multiple transcript variants, each encoding a distinct protein isoform, have been identified.Especificaciones
GSTM4 | |
Polyclonal | |
Unconjugated | |
GSTM4 | |
glutathione S transferase mu 4, GST class mu 4, GST Mu2, GSTM4, GSTM4 4, GTM4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14865, 2948, 499689 | |
-20°C | |
Liquid |
Immunofluorescence, Western Blot, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q03013 | |
Gstm4 | |
GSTM4 Fusion Protein Ag10340 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |