missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSTA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 14475-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Cytosolic and membrane-bound forms of glutathione S-transferase are encoded by two distinct supergene families. These enzymes function in the detoxification of electrophilic compounds, including carcinogens, therapeutic drugs, environmental toxins and products of oxidative stress, by conjugation with glutathione. The genes encoding these enzymes are known to be highly polymorphic. These genetic variations can change an individuals susceptibility to carcinogens and toxins as well as affect the toxicity and efficacy of some drugs. At present, eight distinct classes of the soluble cytoplasmic mammalian glutathione S-transferases have been identified: alpha, kappa, mu, omega, pi, sigma, theta and zeta. This gene encodes a glutathione S-tranferase belonging to the alpha class. The alpha class genes, located in a cluster mapped to chromosome 6, are the most abundantly expressed glutathione S-transferases in liver. In addition to metabolizing bilirubin and certain anti-cancer drugs in the liver, the alpha class of these enzymes exhibit glutathione peroxidase activity thereby protecting the cells from reactive oxygen species and the products of peroxidation.Especificaciones
GSTA1 | |
Polyclonal | |
Unconjugated | |
GSTA1 | |
Glutathione S transferase A1, GST class alpha member 1, GST epsilon, GST HA subunit 1, GST2, GSTA1, GSTA1 1, GTH1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14857, 24421, 2938 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P00502, P08263, P13745 | |
Gsta1 | |
GSTA1 Fusion Protein Ag5849 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |