missing translation for 'onlineSavingsMsg'
Learn More

GST Tag Mouse anti-Tag, Clone: 3G12B10, Proteintech

Mouse Monoclonal Antibody

Marca:  Proteintech 66001-2-IG-150UL

Detalles adicionales : Peso : 0.00010kg

Código de producto. 16817495

  • 189.00€ / 150 microlitros
Fecha estimada de envío: 20-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This antibody recognizes the GST-tag fused to either the N- or C-terminus of targeted proteins.

GST (Glutathione S-transferase) is a 26 kDa protein, present in eukaryotes and in prokaryotes, where they catalyze a variety of reactions. Glutathione Stransferase is used to create the GST gene fusion system. This GST-fusion protein can then be purified from cells via its high affinity for glutathione. Fusion Proteins offer an important biological assay for direct protein-to-protein interactions. The GST gene fusion system has been introduced into numerous expression vectors.
Especificaciones

Especificaciones

GST Tag
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Mouse
150 μL
Primary
Antibody
IgG2b
Immunoprecipitation, Western Blot
3G12B10
Unconjugated
GST, GST Tag
Protein A
RUO
-20°C
Liquid
Tag
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales