missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRHPR Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 51013-2-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes an enzyme with hydroxypyruvate reductase, glyoxylate reductase, and D-glycerate dehydrogenase enzymatic activities. The enzyme has widespread tissue expression and has a role in metabolism. Type II hyperoxaluria is caused by mutations in this gene.Especificaciones
GRHPR | |
Polyclonal | |
Unconjugated | |
GRHPR | |
GLXR, GLYD, GRHPR, PH2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
76238, 9380 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q91Z53, Q9UBQ7 | |
GRHPR | |
GRHPR Fusion Protein Ag0484 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |