Learn More
Fisherbrand™ Grade 1105 Cellulose Technical Filter Paper
Suitable for filtration of large volume of liquid.
Marca: Fisherbrand™
Detalles adicionales : Peso : 0.01000kg
Descripción
- Very thick smooth paper
- Wet strengthened
- With high separation capacity
Especificaciones
Disc | |
3 to 6 μm | |
For dyes, Ether, Alcoholic solutions, Eaux de Cologne, salted solutions, vegetal extracts, hair solutions, perfumes, tinctures, saccharine juices, vinegar | |
100 Pack | |
185 mm |
Cellulose | |
3 to 6 μm | |
1105 | |
0.39 mm | |
185 mm |