Learn More
Fisherbrand™ Grade 1100 Cellulose Technical Filter Paper
For filtration of coarse particles and gelatinous precipitates.
Marca: Fisherbrand™
Detalles adicionales : Peso : 0.01000kg
Descripción
- Creped thin paper
- Wet strengthened
- For filtering viscous liquids as arabic gum, gelatine, emulsions like agar-agar, alginate
Especificaciones
Disc | |
50 to 60 μm | |
Viscous liquids as Arabic gum, Gelatin, emulsions like Agar-agar, Alginate, protective Paper for filter presses | |
100 Pack | |
110 mm |
Cellulose | |
50 to 60 μm | |
1100 | |
0.2 mm | |
110 mm |